Learn More
Thermco™ Infrared Thermometer
Wall mount infrared thermometer
Manufacturer:  Thermco™ ACCD0973IR
Catalog No. 04-500-332
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
This item is not returnable. View return policy
Description
Description
  • Wall mount
  • Hands-free/No human-to-human contact
  • Audible and visual alarms
  • Can be mounted on tripod
  • Selectable °C or °F
  • Includes USB wall charger
Specifications
Specifications
122°F
32°F
CE/FCC/RoHS
Temperature Scanning
170 mm
ABS Plastic
ACCUTHERM™
24 oz.
5.5 in.
±1.0°F/±0.2°C
50°C
0°C
0.9
6.7 in.
DC 5 V USB Wall Charger
0.5 seconds
0.1
350 g
115 mm
18650 Li-Ion Rechargeable (Not Included)