Learn More
Abnova™ ING1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00003621T02
Catalog No. 89-949-032
Add to cart
Specifications
Specifications
293T
NM_198219
pCMV-ING1 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
ING1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 31.9kDa
Transient overexpression cell lysate was tested with anti-ING1 antibody (Mfr. No. H00003621-D01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human