Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ING1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550511

Catalog No. PA550511


Add to cart

Description

Description

The antibody detects endogenous levels of total ING1 protein.

This gene encodes a tumor suppressor protein that can induce cell growth arrest and apoptosis. The encoded protein is a nuclear protein that physically interacts with the tumor suppressor protein TP53 and is a component of the p53 signaling pathway. Reduced expression and rearrangement of this gene have been detected in various cancers. Multiple alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications

Specifications

ING1
Polyclonal
Unconjugated
ING1
Liquid
ING1
Synthetic peptide corresponding to residues near the N terminal of human inhibitor of growth family, member 1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1.6 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q9UK53, Q9QXV3
p33, p47, p33ING1, p24ING1c, p33ING1b, p47ING1a
Rabbit
IgG
100 μL
-20°C
Primary
26356, 3621
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit