Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ING4 Goat anti-Human, DyLight 680, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001682FR
Description
ING4 Polyclonal specifically detects ING4 in Human samples. It is validated for Western Blot, ELISA.Specifications
ING4 | |
Polyclonal | |
Western Blot, ELISA | |
candidate tumor suppressor p33 ING1 homolog, inhibitor of growth family, member 4, inhibitor of growth protein 4, MGC12557, my036, p29ING4brain my036 protein | |
This affinity purified ING4 Antibody was prepared from whole goat serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near aa 150-175 of Human ING4 protein (Inhibitor of growth family, member 4). This sequence only shows homology to isoform 1 for ING4. (Uniprot: Q9UNL4) | |
0.1 mL | |
Chromatin Research, Tumor Suppressors | |
51147 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
DyLight 680 | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title