Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ING4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17938320UL

 View more versions of this product

Catalog No. NBP17938320


Add to cart

Description

Description

ING4 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ING4
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
ING4
Synthetic peptide directed towards the middle region of human ING4The immunogen for this antibody is ING4. Peptide sequence DLKEKQIESSDYDSSSSKGKKSRTQKEKKAARARSKGKNSDEEAPKTAQK.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1:1000
NP_057246
candidate tumor suppressor p33 ING1 homolog, inhibitor of growth family, member 4, inhibitor of growth protein 4, MGC12557, my036, p29ING4brain my036 protein
Rabbit
IgG
20ul
Chromatin Research, Tumor Suppressors
Primary
51147
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit