Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ING4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179383
Description
ING4 Polyclonal specifically detects ING4 in Human samples. It is validated for Western Blot.Specifications
ING4 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
candidate tumor suppressor p33 ING1 homolog, inhibitor of growth family, member 4, inhibitor of growth protein 4, MGC12557, my036, p29ING4brain my036 protein | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Human: 100%; Mouse: 100%; Canine: 86%; Pig: 86%; Rabbit: 86%; Rat: 86%; Chicken: 80%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_057246 | |
ING4 | |
Synthetic peptide directed towards the middle region of human ING4The immunogen for this antibody is ING4. Peptide sequence DLKEKQIESSDYDSSSSKGKKSRTQKEKKAARARSKGKNSDEEAPKTAQK. | |
100 μL | |
Chromatin Research, Tumor Suppressors | |
51147 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction