Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ING4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | ING4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179383
|
Novus Biologicals
NBP179383 |
100 μL |
Each of 1 for $436.00
|
|
Description
ING4 Polyclonal specifically detects ING4 in Human samples. It is validated for Western Blot.Specifications
ING4 | |
Polyclonal | |
Rabbit | |
Chromatin Research, Tumor Suppressors | |
NP_057246 | |
51147 | |
Synthetic peptide directed towards the middle region of human ING4The immunogen for this antibody is ING4. Peptide sequence DLKEKQIESSDYDSSSSKGKKSRTQKEKKAARARSKGKNSDEEAPKTAQK. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
candidate tumor suppressor p33 ING1 homolog, inhibitor of growth family, member 4, inhibitor of growth protein 4, MGC12557, my036, p29ING4brain my036 protein | |
ING4 | |
IgG | |
Affinity Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title