Learn More
Invitrogen™ ING5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596627
Description
Immunogen sequence: MATAMYLEHY LDSIENLPCE LQRNFQLMRE LDQRTEDKKA EIDILAAEYI STVKTLSPDQ RVERLQKIQN AYSKCKEYSD DKVQLAMQTY EMVDKHIRRL DADLARFEAD LKDKMEGSDF ESSGGRGLKK GRGQKEKRGS RGRGRRTSEE DTPKKKKHKG GSEFTDTILS VHPSDVLDMP VDPNEPTYCL CHQVSYGEMI GCDNPDCPIE WFHFACVDLT; Positive Samples: Jurkat, A-549, 293T; Cellular Location: Nucleus.
ING5 is similar to ING1, a tumor suppressor protein that can interact with TP53, inhibit cell growth, and induce apoptosis. This protein contains a PHD-finger, which is a common motif in proteins involved in chromatin remodeling. This protein can bind TP53 and EP300/p300, a component of the histone acetyl transferase complex, suggesting its involvement in TP53-dependent regulatory pathway.
Specifications
ING5 | |
Polyclonal | |
Unconjugated | |
ING5 | |
1700001C14Rik; 1700027H23Rik; 1810018M11Rik; AI225768; ING5; inhibitor of growth family member 5; inhibitor of growth family, member 5; inhibitor of growth protein 5; p28ING5 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
84289 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.78 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8WYH8 | |
ING5 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human ING5 (NP_1157052). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.