Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ING5 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$177.16 - $436.00
Specifications
Antigen | ING5 |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
ING5 Polyclonal specifically detects ING5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ING5 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
inhibitor of growth family, member 5, inhibitor of growth protein 5, p28ING5FLJ23842 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human ING5 (NP_115705.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Tumor Suppressors | |
PBS with 50% glycerol, pH7.3. | |
84289 | |
IgG | |
Affinity purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title