INH1, Tocris™

Hec1 inhibitor; causes arrest of mitosis

Manufacturer: R&D Systems 3551/50

Quantity 50mg

 View more versions of this product

Catalog No. 35-515-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C18H16N2OS·HCl
    Formula Weight 344.86
    Synonym N-[4-(2, 4-Dimethylphenyl)-2-thiazolyl]benzamide hydrochloride
    Chemical Name or Material INH1

    Specifications

    Formula Weight 344.86
    Quantity 50mg
    Assay Percent Range >99%
    Molecular Formula C18H16N2OS·HCl
    CAS 313553-47-8
    Synonym N-[4-(2, 4-Dimethylphenyl)-2-thiazolyl]benzamide hydrochloride
    Recommended Storage Desiccate at +4°C
    Chemical Name or Material INH1

    Desiccate at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit