Learn More
Abnova™ INHBB 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00003625T02
Catalog No. 89-949-059
Add to cart
Description
Description
Specifications
Specifications
293T
BC030029
pCMV-INHBB full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
INHBB 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 45.2kDa
Transient overexpression cell lysate was tested with anti-INHBB antibody (Mfr. No. H00003625-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human