Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INHBE Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551101

Catalog No. PA551101


Add to cart

Description

Description

INHBE Polyclonal Antibody detects endogenous levels of INHBE protein.

Inhibins and activins inhibit and activate, respectively, the secretion of follitropin by the pituitary gland. Inhibins/activins are involved in regulating a number of diverse functions such as hypothalamic and pituitary hormone secretion, gonadal hormone secretion, germ cell development and maturation, erythroid differentiation, insulin secretion, nerve cell survival, embryonic axial development or bone growth, depending on their subunit composition. Inhibins appear to oppose the functions of activins.
Specifications

Specifications

INHBE
Polyclonal
Unconjugated
INHBE
Liquid
INHBE
Synthesized peptide derived from the C-terminal region of human Inhibin B-E.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P58166, O08717, O88959
INHBE, Inhibin beta E chain, Activin beta-E chain
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
16326, 83711, 83729
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit