Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inhibin alpha Mouse anti-Human, Clone: INHA/1168, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Inhibin alpha
Clone INHA/1168
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122340
View Documents
Novus Biologicals
NBP30743320UG
20 μg
Each for $138.37
Add to cart
 
NB122339
View Documents
Novus Biologicals
NBP30743310UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Inhibin alpha Monoclonal antibody specifically detects Inhibin alpha in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Inhibin alpha
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha
Recombinant fragment of human Inhibin alpha protein (around aa 233-362) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
INHA/1168
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Apoptosis, Cancer
1 mM PBS with 0.05% BSA
3623
IgG2b Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit