Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inhibin alpha Mouse anti-Human, Clone: INHA/4265, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Inhibin alpha
Clone INHA/4265
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122341
View Documents
Novus Biologicals
NBP30743410UG
100 μg
Each for $352.89
Add to cart
 
NB122342
View Documents
Novus Biologicals
NBP30743420UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

Inhibin alpha Monoclonal antibody specifically detects Inhibin alpha in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Inhibin alpha
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha
A synthetic peptide (around aa 73-96) of alpha-subunit of human Inhibin alpha protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
INHA/4265
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Apoptosis, Cancer
1 mM PBS with 0.05% BSA
3623
IgG2a
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit