Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Inhibin alpha Mouse anti-Human, Clone: R1, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | Inhibin alpha |
---|---|
Clone | R1 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122392
|
Novus Biologicals
NBP30745920UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
Inhibin alpha Monoclonal specifically detects Inhibin alpha in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Inhibin alpha | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer | |
1 mM PBS with 0.05% BSA | |
3623 | |
IgG2a κ | |
Protein A or G purified |
R1 | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha | |
A synthetic peptide corresponding to Human Inhibin alpha a 233-264. Note that this peptide corresponds to the N-terminus of the mature region of the human inhibin alpha subunit. | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title