Learn More
Inhibin alpha Mouse anti-Human, DyLight 488, Clone: PO12/8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP261936G
Catalog No. NBP261936G
Add to cart
Description
Description
Inhibin alpha Monoclonal antibody specifically detects Inhibin alpha in Human samples. It is validated for ELISA, Immunohistochemistry.
Specifications
Specifications
Inhibin alpha
PO12/8
ELISA, Immunohistochemistry
Purified
INHA
Synthetic peptide corresponding to epitope region aa73-96 of the alphaC region subunit of inhibin A
Protein A purified
Apoptosis, Cancer
Primary
3623
ELISA, Immunohistochemistry
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human