Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Inhibin alpha Mouse anti-Human, DyLight 680, Clone: PO12/8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP261936FR
Description
Inhibin alpha Monoclonal antibody specifically detects Inhibin alpha in Human samples. It is validated for ELISA, Immunohistochemistry.Specifications
Inhibin alpha | |
PO12/8 | |
ELISA, Immunohistochemistry | |
PBS with 0.05% Sodium Azide | |
3623 | |
Mouse | |
IgG2a | |
Primary | |
0.1 ml | |
Store at 4C in the dark. |
ELISA,Immunohistochemistry | |
DyLight 680 | |
Purified | |
A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha | |
INHA | |
Synthetic peptide corresponding to epitope region aa73-96 of the alphaC region subunit of inhibin A | |
Monoclonal | |
Protein A purified | |
Apoptosis, Cancer | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title