Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Inhibin beta A Human Recombinant Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
$344.00 - $976.00
Specifications
Concentration | 1mg/mL |
---|---|
For Use With (Application) | SDS-Page |
Formulation | 20mM Tris 8.0 containing 10% glycerol |
Gene ID (Entrez) | 3624 |
Molecular Weight | TMW: 15.2kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25198601
|
Novus Biologicals™
NBP2519860.1MG |
0.1mg |
Each for $344.00
|
|
NBP25198605
|
Novus Biologicals™
NBP2519860.5MG |
0.5mg |
Each for $976.00
|
|
Description
A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 311-426 of Human Inhibin beta A The Recombinant Human Activin A Protein is derived from E. coli. The Recombinant Human Activin A Protein has been validated for the following applications: SDS-Page.Specifications
1mg/mL | |
20mM Tris 8.0 containing 10% glycerol | |
TMW: 15.2kDa | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
Inhibin beta A | |
Yes |
SDS-Page | |
3624 | |
>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain | |
Activin beta-A chain, beta A (activin A, activin AB alpha polypeptide), EDF, erythroid differentiation factor, Erythroid differentiation protein, follicle-stimulating hormone-releasing protein, FRP, FSH-releasing protein, inhibin beta A chain, inhibin, beta A, Inhibin, beta-1 | |
Unlabeled |
For research use only and is not approved for use in humans or in clinical diagnosis
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title