Learn More
Thermo Scientific LinerGOLD GC Liners
Use the gold-standard LinerGOLD GC Liners for consistently high accuracy, sensitivity, and precision in your gas chromatography (GC) analysis.
Manufacturer: Thermo Scientific 45354030
Description
Use the gold standard in GC liner performance and consistency. Thermo Scientific LinerGOLD GC Liners provide enhanced inertness, leading to increased accuracy, sensitivity, and precision in your gas chromatography (GC) analysis. These injection port liners deliver:
- Increased accuracy and sensitivity in analysis, preventing the loss of sensitive compounds in analysis
- Lower detection limits when analyzing active and sensitive compounds
- Excellent reproducibility gives confidence in your results from liner to liner
- Consistently high level of performance time after time
Specifications
Split Straight Liner | |
25 | |
5 mm | |
105 mm |
Thermo Scientific Instruments | |
None | |
8 mm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.