Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ J50 Series Printer Ink - Yellow
Large J50 series ink cartridges reduce the amount of changeovers or replacements needed. Each color cartridge is separate to optimize ink and allow individual replacement.
Manufacturer: Brady™ J50YL
Description
- Combination of the four CMYK ink cartridges produces 16 million color possibilities
- Ink volume can allow for the printing of thousands of labels before the ink runs out depending on size and ink coverage of the label
- Four separate ink cartridges allow the usage of all of the ink in each cartridge and individual replacement as needed
- Ink cartridge replacement is simple and efficient in less than 10 seconds
Specifications
Not Autoclavable | |
0.94 in. | |
0.94 x 4 x 2.25 in. | |
2.25 in. | |
Ink | |
2.25 x 4 x 0.94 in. |
Yellow | |
Ink | |
Ink | |
Ink Jet | |
1 Ea. | |
4 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title