Learn More
Innocal™ NIST™-Traceable Calibration; for Temperature Meters
NIST™-traceable calibrations for temperature meters
$213.50
Specifications
Product Type Calibration
View More Specs
Products 1
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
15-700-351
View Documents
Innocal™
1700006
Thermistor Meter
Each for $213.50
Add to cart
Description
Description
  • Provides traceability to NIST™
  • Lists the environmental conditions present at time of calibration
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
  • Not an accredited certificate
Specifications
Specifications
Calibration