Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innova™ Laboratory Tube Plug Closure Non-distribution product as customer accommodation.

Made with polyethylene in a choice of colors to color-code tubes

Manufacturer:  Innova™ 6SNCW

 View more versions of this product

Catalog No. 11-387-62


Add to cart

Description

Description

  • For 16mm tubes

Specifications

Specifications

Plug
White
16mm tubes
16mm
Polyethylene
Snap Cap
0.62 in.
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit