Learn More
Novus Biologicals™ InnovaCoat™ GOLD Antibody Labeling Kit, 60nm, Mini
Use simple pipetting operations to covalently attach antibodies, proteins and other biomolecules to ultra-stable InnovaCoat GOLD nanoparticles. Hands-on time is about two minutes and the conjugate is ready within fifteen.
Manufacturer:  Novus Biologicals™ 2320005
 View more versions of this product
Catalog No. 23-200-05
  Check Availability
Add to cart
Alert:
For Research Use Only
Specifications
Description
Features
  • Proprietary surface coat reduces risk of sub-optimal antibody performance
  • Ultra-stable
  • Covalently link antibodies and analytes
  • Simple conjugation processes
  • Different surface chemistries
Benefits
  • Antibody-GOLD conjugates in just 15 minutes
  • Enhanced assay sensitivity
  • Protective coat prevents metal-antibody interactions
  • Coating procedure not dependent on isoelectric point of antibody
  • Rapid screening of multiple antibodies for assay development
  • Reduced antibody loading without aggregation
  • No pH titrations
  • Fully scalable – from R&D to production
  • Custom InnovaCoat™ labeling services
Description & Specifications
Specifications
3 Reactions
Store at 4°C. Do not freeze.
Antibody Labeling Kit