Novus Biologicals™ InnovaCoat™ GOLD Antibody Labeling Kit, 60nm, Mini
Use simple pipetting operations to covalently attach antibodies, proteins and other biomolecules to ultra-stable InnovaCoat GOLD nanoparticles. Hands-on time is about two minutes and the conjugate is ready within fifteen.
Manufacturer: Novus Biologicals™ 2320010
 View more versions of this product
Catalog No. 23-200-10
  Check Availability
Add to Cart
Alert
For Research Use Only
    Specifications
    Product Type Antibody Labeling Kit
    Quantity 10 Reactions
    Storage Requirements Store at 4°C. Do not freeze.
    Features
    • Proprietary surface coat reduces risk of sub-optimal antibody performance
    • Ultra-stable
    • Covalently link antibodies and analytes
    • Simple conjugation processes
    • Different surface chemistries
    Benefits
    • Antibody-GOLD conjugates in just 15 minutes
    • Enhanced assay sensitivity
    • Protective coat prevents metal-antibody interactions
    • Coating procedure not dependent on isoelectric point of antibody
    • Rapid screening of multiple antibodies for assay development
    • Reduced antibody loading without aggregation
    • No pH titrations
    • Fully scalable – from R&D to production
    • Custom InnovaCoat™ labeling services
  • Description & Specifications
Related Products
Product Content Feedback