Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ InnovaCoat™ GOLD Goat Anti-Mouse IgG Gold (10OD) Antibody Labeling Kit, 40nm

Unique coated nanoparticle with a strongly bound protective surface coat

$199.00 - $429.00

Specifications

Storage Requirements Store at 4C. Do not freeze.
Target InnovaCoat GOLD - 40nm Goat Anti-Mouse IgG Gold (10OD)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
2160200 View Documents Novus Biologicals™
2160200
0.2mL Each for $199.00
Add to cart
 
2161000 View Documents Novus Biologicals™
2161000
1mL Each for $429.00
Add to cart
 
Specifications

Specifications

Store at 4C. Do not freeze.
InnovaCoat GOLD - 40nm Goat Anti-Mouse IgG Gold (10OD)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit