Learn More
Novus Biologicals™ InnovaCoat™ GOLD Hydrazide Antibody Labeling Kit, 40nm
Quick and easy attachment of antibodies. Hydrazide groups bind antibodies via the Fc region, orienting the two antigen-binding arms in space.
Manufacturer:  Novus Biologicals™ 2800300
 View more versions of this product
Catalog No. 28-003-00
Add to cart
Alert:
For Research Use Only
Description
Description
Features
  • Proprietary surface coat
  • Ultra-stable
  • Covalently link antibodies and analytes
  • Simple conjugation processes
  • Different surface chemistries
Benefits
  • Enhanced assay sensitivity
  • Protective coat prevents metal-antibody interactions
  • Rapid screening of multiple antibodies for assay development
  • Reduced antibody loading without aggregation
  • No pH titrations
  • Fully scalable – from R&D to production
  • Custom InnovaCoat™ labeling services
Specifications
Specifications
1 x 30-90μg Reaction
Store at 4°C. Do not freeze.
Antibody Labeling Kit