Novus Biologicals™ InnovaCoat™ GOLD Hydrazide Antibody Labeling Kit, 40nm
Quick and easy attachment of antibodies. Hydrazide groups bind antibodies via the Fc region, orienting the two antigen-binding arms in space.
Manufacturer: Novus Biologicals™ 2800000
 View more versions of this product
Catalog No. 28-000-00
  Check Availability
Add to Cart
Alert
For Research Use Only
    Specifications
    Product Type Antibody Labeling Kit
    Quantity 3 x 10-30μg Reaction
    Storage Requirements Store at 4°C. Do not freeze.
    Features
    • Proprietary surface coat
    • Ultra-stable
    • Covalently link antibodies and analytes
    • Simple conjugation processes
    • Different surface chemistries
    Benefits
    • Enhanced assay sensitivity
    • Protective coat prevents metal-antibody interactions
    • Rapid screening of multiple antibodies for assay development
    • Reduced antibody loading without aggregation
    • No pH titrations
    • Fully scalable – from R&D to production
    • Custom InnovaCoat™ labeling services
  • Description & Specifications
Related Products
Product Content Feedback