Novus Biologicals™ InnovaGOLD Gold Nanoparticles Antibody Labeling Kit, 40nm, 1 OD

Manufacturer: Novus Biologicals™ 2010010

 View more versions of this product

Catalog No. 20-100-10

  Check Availability
Add to Cart

Alert

For Research Use Only


    Specifications

    Product Type Antibody Labeling Kit
    Quantity 10mL
    Storage Requirements Store at 4°C. Do not freeze.

    • Uniform Shape
    • Low co-efficient of variation in terms of size
    • High efficiency antibody binding for maximum sensitivity
    • Available at high concentrations to simplify assay development

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit