Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ 40nm Colloidal Gold Nanoparticles 10 OD
Supplier: Novus Biologicals™ 2100010
Alert:
For Research Use Only
Description
- Uniform Shape
- Low co-efficient of variation in terms of size
- High efficiency antibody binding for maximum sensitivity
- Available at high concentrations to simplify assay development
Specifications
Store at 4°C. Do not freeze. | |
10 mL | |
40nm Colloidal Gold Nanoparticles 10 OD | |
Antibody Labeling Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction