Learn More
Innovating Science™ LyoQuick Freeze Dried Bacteria Cultures Products for Science Education
Easy to use lyophillized bacterial cultures for use in classroom for educational purposes. Innovating Science™ LyoQuick Freeze Dried Bacteria Cultures, eliminates several steps required for rehydration of freeze-dried cultures, thus reducing the risk of contamination.
Manufacturer:  Innovating Science™ IS3603
 View more versions of this product
Catalog No. S29045
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Each tube contains two ampules, one containing freeze-dried bacteria and one self-contained sterile rehydration broth
  • To use, pinch tube to break internal ampules and let freezedried organisms rehydrate, then dispense rehydrated organisms into broth or onto agar through dropper tip
  • Available for seven different types of bacteria
Specifications
Specifications
White
LyoQuick Freeze Dried Bacteria
1
20
13
Format: Freeze Dried
Micrococcus luteus
4 x 0.5 x 0.5 in.
14
7