Learn More
Innovating Science™ Make Your Own Bromine Water Educational Kit Products for Science Education
To prepare fresh bromine water in a couple of minutes. Innovating Science Make Your Own Bromine Water Educational Kit, helps save on hazardous transport costs and avoid storing elemental bromine in storage room.
Manufacturer:  Innovating Science™ IS5023
Catalog No. S28497
Add to cart
Alert:
WARNING: This product can expose you to chemicals including Potassium Bromate and Strong Inorganic Acid Mists containing sulfuric acid, which are known to the State of California to cause cancer.
Includes: 2.15g Potassium Bromide; 0.6g  Potassium Bromate; 25mL Sulfuric Acid; 50mL Bromine Water-Btl/Label Only
Description
Description
Helps students to prepare bromine water instantly for experiments
Specifications
Specifications
White
Make Your Own Bromine Water Educational Kit
4.25 x 4.625 x 5.5 in.
14
7
Educational use
1
20
13