Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Physical and Chemical Properties of Matter Educational Kit
SCI_ED

To perform a series of experiments focusing on the physical properties of density, smell, color, melting point, boiling point and solubility. Innovating Science™ Physical and Chemical Properties of Matter Educational Kit focuses on developing a better understanding of what is matter and what the different properties are.

Manufacturer:  Aldon Corporation IS2517

Catalog No. S28489


Add to cart

Alert:

WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid and Respirable Crystalline Silica, which are known to the State of California to cause cancer.


Description

Description

  • Contains enough materials for 10 groups
  • Teacher's manual and student Study Guide copymasters are included
  • Kit Includes:
    • 0.1M Cupric Sulfate (10 x 10 mL)
    • 0.1M Sodium Hydroxide (10 x 10 mL)
    • 0.1M Hydrochloric Acid (10 x 10 mL)
    • 0.1M Sodium Carbonate (10 x 10 mL)
    • 1.0M Ammonium Hydroxide (10 x 10 mL)
    • 1.0M Acetic Acid (10 x 10 mL)
    • Unknown (10 x 3 mL)
    • 30 g Sodium Chloride
    • 30 g Calcium Chloride
    • 30 g Dextrose
    • 30 g Sand
    • 30 g Talc Powder
    • Zinc Sample (10 x 5 g)
    • Copper Sample (10 x 5 g)
    • Aluminum Sample (10 x 2 g)
    • Glass Sample (10 x 2 g)
    • 1 pack pH Strips
    • 10 Preprinted Acetate Sheets
    • 10 Thermometers
Specifications

Specifications

1 Kit
13
14
White
10 groups
7
20
12.125 x 9.25 x 6.5 in.
Physical and Chemical Properties of Matter Educational Kit
Educational use
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit