Learn More
Innovative Med Tech Plastic Blood Gas Capillary Tubes: Additives
Additives for Innovative Med Tech Plastic Blood Gas Capillary Tubes
Manufacturer:  INNOVATIVE MEDICAL TECHNOLOGIES INC 102180
 View more versions of this product
Catalog No. 22-757-146
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Tube
Blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening
FDA; OSHA; CDC
180mm
280μL
Plastic
Balanced Li-Heparin
2.35mm