Innovative Med Tech Plastic Blood Gas Capillary Tubes: Additives
Additives for Innovative Med Tech Plastic Blood Gas Capillary Tubes
Manufacturer: INNOVATIVE MEDICAL TECHNOLOGIES INC 101150
 View more versions of this product
Catalog No. 22-757-137
  Check Availability
Add to Cart
    Specifications
    Diameter (Metric) Inner 1.75mm
    Length (Metric) Tube 150mm
    Product Type Tube
    Applications Blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening
    Capacity (Metric) 165μL
    Certifications/Compliance FDA; OSHA; CDC
    Additive Balanced Li-Heparin
    Material Plastic
  • Description & Specifications
Related Products
Product Content Feedback