Innovative Med Tech Plastic Blood Gas Capillary Tubes: Additives
Additives for Innovative Med Tech Plastic Blood Gas Capillary Tubes
Manufacturer: INNOVATIVE MEDICAL TECHNOLOGIES INC 101180
 View more versions of this product
Catalog No. 22-757-138
  Check Availability
Add to Cart
    Specifications
    Product Type Tube
    Material Plastic
    Capacity (Metric) 200μL
    Certifications/Compliance FDA; OSHA; CDC
    Additive Balanced Li-Heparin
    Diameter (Metric) Inner 180mm
    Length (Metric) Tube 1.75mm
    Applications Blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening
  • Description & Specifications
Related Products
Product Content Feedback