Learn More
Innovative Med Tech Plastic Capillary Tubes
Replace glass or Mylar™-coated Natelson and Caraway tubes
Manufacturer:  Innovative Med Tech 100110
 View more versions of this product
Catalog No. 22-757-122
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Replace glass or Mylar™-coated Natelson and Caraway tubes
  • Made from 100% plastic
  • Use for blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening
Compliance
Meets FDA, OSHA, and CDC mandates
Specifications
Specifications
Tube
10μL
Plastic
30mm
EDTA
Blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening
FDA; OSHA; CDC