Innovative Med Tech Plastic Capillary Tubes
Replace glass or Mylar™-coated Natelson and Caraway tubes
Manufacturer: INNOVATIVE MEDICAL TECHNOLOGIES INC 100130
 View more versions of this product
Catalog No. 22-757-125
  Check Availability
Add to Cart
    Specifications
    Product Type Tube
    Capacity (Metric) 30μL
    Certifications/Compliance FDA; OSHA; CDC
    Additive EDTA
    Length (Metric) Tube 30mm
    Applications Blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening
    Material Plastic
  • Description & Specifications
Related Products
Product Content Feedback