Learn More
Innovative Med Tech Plastic Capillary Tubes
Replace glass or Mylar™-coated Natelson and Caraway tubes
Manufacturer:  INNOVATIVE MEDICAL TECHNOLOGIES INC 100025
 View more versions of this product
Catalog No. 22-757-118
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Tube
Blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening
FDA; OSHA; CDC
30mm
25μL
Plastic
Sodium Heparin