Learn More
Innovative Med Tech Plastic Capillary Tubes
Replace glass or Mylar™-coated Natelson and Caraway tubes
Manufacturer:  INNOVATIVE MEDICAL TECHNOLOGIES INC 100040
 View more versions of this product
Catalog No. 22-757-120
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Tube
Blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening
FDA; OSHA; CDC
30mm
40μL
Plastic
Sodium Heparin