Innovative Med Tech Plastic Hematocrit Capillary Tubes
Replace glass or Mylar™-coated Natelson and Caraway tubes
Manufacturer: INNOVATIVE MEDICAL TECHNOLOGIES INC 103050
 View more versions of this product
Catalog No. 22-757-130
  Check Availability
Add to Cart
    Specifications
    Product Type Tube
    Length (Metric) Tube 50mm
    Certifications/Compliance FDA, OSHA, CDC
    Additive None
    Applications Blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening
    Material Plastic
  • Description & Specifications
Related Products
Product Content Feedback