Learn More
Innovative Med Tech Plastic Hematocrit Capillary Tubes
Replace glass or Mylar™-coated Natelson and Caraway tubes
Manufacturer:  INNOVATIVE MEDICAL TECHNOLOGIES INC 103150
 View more versions of this product
Catalog No. 22-757-131
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Tube
Plastic
Heparin
Blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening
FDA, OSHA, CDC
50mm