Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inorganic Carbon Standard, Reagents

Catalog No. 01335581
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
4 L
Catalog No. Quantity
01-335-581 500 mL
01-335-584 125 mL
01-335-583 1 L
01-335-582 4 L
4 options

Catalog No. 01-335-581

Supplier: Reagents Holdings Llc CC205800500A

Only null left
Add to Cart
Add to Cart

Inorganic Carbon Standard, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Inorganic Carbon Standard
Packaging Poly Bottle
CAS 7732-18-5, 497-19-8
Specific Gravity 1.0045
Physical Form Liquid
Color Colorless
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Industry Type Water Analysis
Quantity 500 mL
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.