Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ iNOS Knockout HeLa Cell Lysate
This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Manufacturer: Novus Biologicals™ NBP265101
Description
The iNOS Knockout HeLa Cell Lysate is derived from human. It has been validated for the following applications: Western Blot.Specifications
LYOPH | |
Western Blot | |
2 Vials | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
iNOS | |
Human | |
0.1 mg cell homogenate lyophilized in RIPA buffer made with double-knockout cell lines |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title