Alfa Aesar™ Inosine-5'-monophosphoric acid disodium salt hydrate, 99%

Manufacturer: Alfa Aesar™ A10729-06

Quantity 5g

 View more versions of this product

Catalog No. AAA1072906

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 352195-40-5
    Molecular Formula C10H11N4Na2O8P·xH2O
    Formula Weight 392.24 (Anhydrous)
    MDL Number MFCD00036201
    Chemical Name or Material Inosine-5'-monophosphoric acid disodium salt hydrate

    Specifications

    Formula Weight 392.24 (Anhydrous)
    TSCA TSCA
    Molecular Formula C10H11N4Na2O8P·xH2O
    MDL Number MFCD00036201
    CAS 352195-40-5
    Quantity 5g
    Recommended Storage 2°C to 8°C
    RTECS Number NM7519500
    Chemical Name or Material Inosine-5'-monophosphoric acid disodium salt hydrate
    Assay 99%

    2°C to 8°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit