Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Inosine-5'-monophosphoric acid disodium salt hydrate, 99%

Manufacturer:  Alfa Aesar™ A1072914

Quantity 25g

 View more versions of this product

Catalog No. AAA1072914

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

352195-40-5
392.24 (Anhydrous)
Inosine-5'-monophosphoric acid disodium salt hydrate
C10H11N4Na2O8P·xH2O
MFCD00036201

Specifications

352195-40-5
392.24 (Anhydrous)
25g
99%
C10H11N4Na2O8P·xH2O
MFCD00036201
Inosine-5'-monophosphoric acid disodium salt hydrate

Safety and Handling

RTECSNumber : NM7519500

TSCA : TSCA

Recommended Storage : 2°C to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit