Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inosine, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
250g
Packaging:
Glass bottle
58-63-9
C10H12N4O5
268.22
MFCD00066770
UGQMRVRMYYASKQ-KQYNXXCUSA-N
inosine,hypoxanthosine,ribonosine,atorel,oxiamin,trophicardyl,selfer,pantholic-l,panholic-l,hypoxanthine riboside
6021
CHEBI:17596
9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
C1=NC(=O)C2=C(N1)N(C=N2)C3C(C(C(O3)CO)O)O
58-63-9
C10H12N4O5
268.22
MFCD00066770
UGQMRVRMYYASKQ-KQYNXXCUSA-N
inosine,hypoxanthosine,ribonosine,atorel,oxiamin,trophicardyl,selfer,pantholic-l,panholic-l,hypoxanthine riboside
6021
CHEBI:17596
9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
C1=NC(=O)C2=C(N1)N(C=N2)C3C(C(C(O3)CO)O)O
Melting Point 212°C to 213°C
Specific Rotation -49.2
Color White
Specific Rotation Condition − 49.20 (18.00°C c=1,H2O)
Quantity 250g
Loss on Drying 1% max.
Packaging Glass bottle
Infrared Spectrum Authentic
Sulfated Ash 0.1% max.
Spectrophotometry 98 to 102% (lambda max. 248.5nm, pH 7), molar absorptivity 12200 +/- 2%, on dry substance
Transmittance 98% min. (c=1, H2O, 430nm, 1cm cell)
Absorbance 1.65 to 1.77 (A250/A260), 0.22 to 0.26 (A280/A260)
Assay Percent Range 99%
Beilstein 31, 25
Merck Index 15, 5018
Solubility Information Solubility in water: 2.1g/100 mL (20°C). Other solubilities: soluble in ethanol
Formula Weight 268.22
Physical Form Crystalline Powder
Percent Purity ≥98.5% (on dry substance)
Chemical Name or Material Inosine
Show More Show Less