Learn More
BioVision IncSupplier Diversity Partner Inosine Fluorometric Assay Kit: Inosine Assay Kit: Fluorometric Assay to Quantify Inosine in a variety of Biological Samples such as Tissue, Cells etc. within 40 min. Rapid, Convenient & Sensitive. Detection Limit: 100 pmol. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K712100
Inosine is commonly found in tRNAs and is essential for proper translation of the genetic code in wobble base pairs. Knowledge of inosine metabolism has led to advances in immunotherapy in recent decades. Inosine and hypoxanthine are also potential markers of ATP catabolic-by-products from oxidative stress which follows acute cardiac ischemia. In BioVision’s Inosine Assay Kit, inosine is converted to hypoxanthine, which will react with the substrate mix and PicoProbe™ to generate fluorescence (Ex/Em = 535/587 nm) in presence of converter and developer enzymes. This assay has a detection limit of of approximately 100 pmol inosine/well. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9855