Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inositol, FCC, 97%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
87-89-8
C6H12O6
180.16
CDAISMWEOUEBRE-UHFFFAOYSA-N
cyclohexane-1,2,3,4,5,6-hexol
OC1C(O)C(O)C(O)C(O)C1O
87-89-8
C6H12O6
180.16
CDAISMWEOUEBRE-UHFFFAOYSA-N
cyclohexane-1,2,3,4,5,6-hexol
OC1C(O)C(O)C(O)C(O)C1O

I1004, 87-89-8, C6H12O6

Spectrum™ Inositol, FCC is a vitamin-like substance, sometimes known colloquially as ′vitamin B-8′, is a beneficial nutrient that has been implicated in the treatment of some behavioral or emotional disorders. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Bottle
Ignition Residue 0.001
Loss on Drying 0.005
Melting Range 224-227 C
Quantity 500 g
CAS Max % 1
Percent Purity 97%