Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Inositol Monophosphatase 3/IMPAD1 Human Recombinant Protein

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

Supplier:  Novus Biologicals™ NBP2519930.05MG

 View more versions of this product

Catalog No. NBP25199305


Add to cart

Description

Description

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 34-359 of Human Inositol Monophosphatase 3/IMPAD1 The Recombinant Human Inositol Monophosphatase 3/IMPAD1 Protein is derived from E. coli. The Recombinant Human Inositol Monophosphatase 3/IMPAD1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications

Specifications

0.25mg/mL
Phosphate buffered saline (pH7.4)
TMW: 37.6kDa
0.05mg
EC 3.1.3, EC 3.1.3.25, FLJ20421, IMP 3, IMPA3IMPase 3, inositol monophosphatase 3, inositol monophosphatase domain containing 1, Inositol monophosphatase domain-containing protein 1, Inositol-1(or 4)-monophosphatase 3, Myo-inositol monophosphatase A3
Specific acitivty > 3300 pmole/min/ug, its ability to dephosphorylate adenosine 3'5'-diphosphate sodium slat at pH 7.5, 25C.
Yes
SDS-Page
54928
>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Inositol Monophosphatase 3/IMPAD1
Unlabeled
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For research use only and is not approved for use in humans or in clinical diagnosis