Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Inositol Monophosphatase 3/IMPAD1 Human Recombinant Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity
$344.00 - $976.00
Specifications
Concentration | 0.25mg/mL |
---|---|
For Use With (Application) | SDS-Page |
Formulation | Phosphate buffered saline (pH7.4) |
Gene ID (Entrez) | 54928 |
Molecular Weight | TMW: 37.6kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25993001
|
Novus Biologicals™
NBP2519930.01MG |
0.01mg |
Each for $344.00
|
|
NBP25199305
|
Novus Biologicals™
NBP2519930.05MG |
0.05mg |
Each for $976.00
|
|
Description
A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 34-359 of Human Inositol Monophosphatase 3/IMPAD1 The Recombinant Human Inositol Monophosphatase 3/IMPAD1 Protein is derived from E. coli. The Recombinant Human Inositol Monophosphatase 3/IMPAD1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.Specifications
0.25mg/mL | |
Phosphate buffered saline (pH7.4) | |
TMW: 37.6kDa | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
Inositol Monophosphatase 3/IMPAD1 | |
Unlabeled |
SDS-Page | |
54928 | |
>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain | |
EC 3.1.3, EC 3.1.3.25, FLJ20421, IMP 3, IMPA3IMPase 3, inositol monophosphatase 3, inositol monophosphatase domain containing 1, Inositol monophosphatase domain-containing protein 1, Inositol-1(or 4)-monophosphatase 3, Myo-inositol monophosphatase A3 | |
Specific acitivty > 3300 pmole/min/ug, its ability to dephosphorylate adenosine 3'5'-diphosphate sodium slat at pH 7.5, 25C. | |
Yes |
For research use only and is not approved for use in humans or in clinical diagnosis
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title